RIE156320778 Rieke Metals, Inc.

4-Chloro-4'-iodo-3-methylbenzophenone

  • MDL number MFCD09801516
  • Molecular Weight : 356.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5189-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5189-22
Availability details for
Quantity
  • Availability
  • Add to Cart