RIE156320781 Rieke Metals, Inc.

4-Chloro-3',5'-difluoro-3-methylbenzophenone

  • MDL number MFCD09801519
  • Molecular Weight : 266.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5189-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5189-42
Availability details for
Quantity
  • Availability
  • Add to Cart