RIE156320787 Rieke Metals, Inc.

3,4-Dichloro-4'-iodobenzophenone

  • MDL number MFCD07775735
  • Molecular Weight : 377.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dichlorophenyl)(4-iodophenyl)methanone
Inchi Code 1S/C13H7Cl2IO/c14-11-6-3-9(7-12(11)15)13(17)8-1-4-10(16)5-2-8/h1-7H
Inchi Key CNQAFPGAWSQBNP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5190-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5190-22
Availability details for
Quantity
  • Availability
  • Add to Cart