RIE156320791 Rieke Metals, Inc.

3,5-Dichloro-4'-iodobenzophenone

  • MDL number MFCD07775736
  • Molecular Weight : 377.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5191-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5191-22
Availability details for
Quantity
  • Availability
  • Add to Cart