RIE156320793 Rieke Metals, Inc.

3,4-Difluoro-2'-iodobenzophenone

  • MDL number MFCD07775697
  • Molecular Weight : 344.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5192-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5192-20
Availability details for
Quantity
  • Availability
  • Add to Cart