RIE156320798 Rieke Metals, Inc.

3,5-Difluoro-3'-iodobenzophenone

  • MDL number MFCD09801527
  • Molecular Weight : 344.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5193-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5193-21
Availability details for
Quantity
  • Availability
  • Add to Cart