RIE156320813 Rieke Metals, Inc.

3,4-Dichloro-3',5'-dimethoxybenzophenone

  • MDL number MFCD09801543
  • Molecular Weight : 311.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dichlorophenyl)(3,5-dimethoxyphenyl)methanone
Inchi Code 1S/C15H12Cl2O3/c1-19-11-5-10(6-12(8-11)20-2)15(18)9-3-4-13(16)14(17)7-9/h3-8H,1-2H3
Inchi Key FWSZRJDEGVMIOR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5195-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5195-47
Availability details for
Quantity
  • Availability
  • Add to Cart