RIE156320815 Rieke Metals, Inc.

2-Chloro-3',4'-dimethylbenzophenone

  • MDL number MFCD09801545
  • Cas Number : 34701-99-0
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)(3,4-dimethylphenyl)methanone
Inchi Code 1S/C15H13ClO/c1-10-7-8-12(9-11(10)2)15(17)13-5-3-4-6-14(13)16/h3-9H,1-2H3
Inchi Key MHSUZDCWVXTKIE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5196-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5196-14
Availability details for
Quantity
  • Availability
  • Add to Cart