RIE156320816 Rieke Metals, Inc.

2-Bromo-3',4'-dimethylbenzophenone

  • MDL number MFCD09801546
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5196-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5196-17
Availability details for
Quantity
  • Availability
  • Add to Cart