RIE156320818 Rieke Metals, Inc.

3,4-Dimethyl-3'-iodobenzophenone

  • MDL number MFCD07775721
  • Molecular Weight : 336.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethylphenyl)(3-iodophenyl)methanone
Inchi Code 1S/C15H13IO/c1-10-6-7-13(8-11(10)2)15(17)12-4-3-5-14(16)9-12/h3-9H,1-2H3
Inchi Key XVECZUFLEFJGAK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5196-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5196-21
Availability details for
Quantity
  • Availability
  • Add to Cart