RIE156320820 Rieke Metals, Inc.

3,5-Dichloro-3',4'-dimethylbenzophenone

  • MDL number MFCD09801548
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5196-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5196-48
Availability details for
Quantity
  • Availability
  • Add to Cart