RIE156320821 Rieke Metals, Inc.

2-Chloro-3',5'-dimethylbenzophenone

  • MDL number MFCD09801549
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5197-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5197-14
Availability details for
Quantity
  • Availability
  • Add to Cart