RIE156320823 Rieke Metals, Inc.

3,5-Dimethyl-2'-iodobenzophenone

  • MDL number MFCD07775704
  • Molecular Weight : 336.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dimethylphenyl)(2-iodophenyl)methanone
Inchi Code 1S/C15H13IO/c1-10-7-11(2)9-12(8-10)15(17)13-5-3-4-6-14(13)16/h3-9H,1-2H3
Inchi Key HCLAHNISVGHQFC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5197-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5197-20
Availability details for
Quantity
  • Availability
  • Add to Cart