RIE156320825 Rieke Metals, Inc.

3,5-Dimethyl-4'-iodobenzophenone

  • MDL number MFCD07775740
  • Molecular Weight : 336.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5197-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5197-22
Availability details for
Quantity
  • Availability
  • Add to Cart