RIE156320833 Rieke Metals, Inc.

4-tert-Butyl-3',5'-dimethyl-4'-methoxybenzophenone

  • MDL number MFCD09801555
  • Molecular Weight : 296.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5198-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5198-34
Availability details for
Quantity
  • Availability
  • Add to Cart