RIE156320841 Rieke Metals, Inc.

4-tert-Butyl-4'-ethoxybenzophenone

  • MDL number MFCD09801563
  • Molecular Weight : 282.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5199-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5199-34
Availability details for
Quantity
  • Availability
  • Add to Cart