RIE156320842 Rieke Metals, Inc.

3,4-Difluoro-4'-ethoxybenzophenone

  • MDL number MFCD09801564
  • Molecular Weight : 262.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5199-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5199-41
Availability details for
Quantity
  • Availability
  • Add to Cart