RIE156320844 Rieke Metals, Inc.

3,4-Dichloro-4'-ethoxybenzophenone

  • MDL number MFCD09801566
  • Cas Number : 106473-07-8
  • Molecular Weight : 295.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5199-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5199-47
Availability details for
Quantity
  • Availability
  • Add to Cart