RIE156320846 Rieke Metals, Inc.

2-Chloro-2'-ethylbenzophenone

  • MDL number MFCD09801568
  • Molecular Weight : 244.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5200-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5200-14
Availability details for
Quantity
  • Availability
  • Add to Cart