RIE156320849 Rieke Metals, Inc.

2-Bromo-2'-ethylbenzophenone

  • MDL number MFCD09801571
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromophenyl)(2-ethylphenyl)methanone
Inchi Code 1S/C15H13BrO/c1-2-11-7-3-4-8-12(11)15(17)13-9-5-6-10-14(13)16/h3-10H,2H2,1H3
Inchi Key MHRSWIKYICMVHB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5200-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5200-17
Availability details for
Quantity
  • Availability
  • Add to Cart