RIE156320851 Rieke Metals, Inc.

4-Bromo-2'-ethylbenzophenone

  • MDL number MFCD09801573
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5200-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5200-19
Availability details for
Quantity
  • Availability
  • Add to Cart