RIE156320853 Rieke Metals, Inc.

2-Ethyl-3'-iodobenzophenone

  • MDL number MFCD09801575
  • Molecular Weight : 336.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5200-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5200-21
Availability details for
Quantity
  • Availability
  • Add to Cart