RIE156320859 Rieke Metals, Inc.

3,5-Dichloro-2'-ethylbenzophenone

  • MDL number MFCD09801581
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5200-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5200-48
Availability details for
Quantity
  • Availability
  • Add to Cart