RIE156320861 Rieke Metals, Inc.

2-Bromo-4'-ethylbenzophenone

  • MDL number MFCD09801583
  • Cas Number : 137327-29-8
  • Molecular Weight : 289.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5201-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5201-17
Availability details for
Quantity
  • Availability
  • Add to Cart