RIE156320866 Rieke Metals, Inc.

3,4-(Ethylenedioxy)-4'-iodobenzophenone

  • MDL number MFCD00517335
  • Molecular Weight : 366.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5202-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-22
Availability details for
Quantity
  • Availability
  • Add to Cart