RIE156320868 Rieke Metals, Inc.

3,5-Difluoro-3',4'-(ethylenedioxy)benzophenone

  • MDL number MFCD09801588
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5202-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-42
Availability details for
Quantity
  • Availability
  • Add to Cart