RIE156320870 Rieke Metals, Inc.

3,5-Dichloro-3',4'-(ethylenedioxy)benzophenone

  • MDL number MFCD09801590
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dichlorophenyl)(2,3-dihydro-1,4-benzodioxin-6-yl)methanone
Inchi Code 1S/C15H10Cl2O3/c16-11-5-10(6-12(17)8-11)15(18)9-1-2-13-14(7-9)20-4-3-19-13/h1-2,5-8H,3-4H2
Inchi Key WXNAZOVMJSPUQH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5202-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5202-48
Availability details for
Quantity
  • Availability
  • Add to Cart