RIE156320873 Rieke Metals, Inc.

4-(Ethylthio)-2'-iodobenzophenone

  • MDL number MFCD07775705
  • Molecular Weight : 368.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(ethylsulfanyl)phenyl](2-iodophenyl)methanone
Inchi Code 1S/C15H13IOS/c1-2-18-12-9-7-11(8-10-12)15(17)13-5-3-4-6-14(13)16/h3-10H,2H2,1H3
Inchi Key BUPXPVQKZSGEPD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5203-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5203-20
Availability details for
Quantity
  • Availability
  • Add to Cart