RIE156320877 Rieke Metals, Inc.

3,5-Dichloro-4'-(ethylthio)benzophenone

  • MDL number MFCD09801594
  • Molecular Weight : 311.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dichlorophenyl)[4-(ethylsulfanyl)phenyl]methanone
Inchi Code 1S/C15H12Cl2OS/c1-2-19-14-5-3-10(4-6-14)15(18)11-7-12(16)9-13(17)8-11/h3-9H,2H2,1H3
Inchi Key AFLVGHZZQXSVGI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5203-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5203-48
Availability details for
Quantity
  • Availability
  • Add to Cart