RIE156320878 Rieke Metals, Inc.

2-Chloro-3'-fluorobenzophenone

  • MDL number MFCD09801595
  • Cas Number : 81029-87-0
  • Molecular Weight : 234.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5204-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5204-14
Availability details for
Quantity
  • Availability
  • Add to Cart