RIE156320881 Rieke Metals, Inc.

3-Bromo-3'-fluorobenzophenone

  • MDL number MFCD09801599
  • Cas Number : 75762-58-2
  • Molecular Weight : 279.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)(3-fluorophenyl)methanone
Inchi Code 1S/C13H8BrFO/c14-11-5-1-3-9(7-11)13(16)10-4-2-6-12(15)8-10/h1-8H
Inchi Key KGAQUGKHKYDPDW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5204-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5204-18
Availability details for
Quantity
  • Availability
  • Add to Cart