RIE156320885 Rieke Metals, Inc.

3,4,4'-Trifluorobenzophenone

  • MDL number MFCD09801608
  • Molecular Weight : 236.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5205-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5205-41
Availability details for
Quantity
  • Availability
  • Add to Cart