RIE156320886 Rieke Metals, Inc.

3,4',5-Trifluorobenzophenone

  • MDL number MFCD09801609
  • Molecular Weight : 236.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)(4-fluorophenyl)methanone
Inchi Code 1S/C13H7F3O/c14-10-3-1-8(2-4-10)13(17)9-5-11(15)7-12(16)6-9/h1-7H
Inchi Key XWWMKJADTYYKPY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5205-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5205-42
Availability details for
Quantity
  • Availability
  • Add to Cart