RIE156320887 Rieke Metals, Inc.

3,5-Dichloro-4'-fluorobenzophenone

  • MDL number MFCD09801611
  • Cas Number : 197439-66-0
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dichlorophenyl)(4-fluorophenyl)methanone
Inchi Code 1S/C13H7Cl2FO/c14-10-5-9(6-11(15)7-10)13(17)8-1-3-12(16)4-2-8/h1-7H
Inchi Key GHIXIBXUOPMWCC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5205-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5205-48
Availability details for
Quantity
  • Availability
  • Add to Cart