RIE156320891 Rieke Metals, Inc.

3'-Fluoro-3-iodo-4'-methoxybenzophenone

  • MDL number MFCD07775708
  • Molecular Weight : 356.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5206-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5206-21
Availability details for
Quantity
  • Availability
  • Add to Cart