RIE156320892 Rieke Metals, Inc.

3'-Fluoro-4-iodo-4'-methoxybenzophenone

  • MDL number MFCD07775727
  • Molecular Weight : 356.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5206-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5206-22
Availability details for
Quantity
  • Availability
  • Add to Cart