RIE156320893 Rieke Metals, Inc.

4-tert-Butyl-3'-fluoro-4'-methoxybenzophenone

  • MDL number MFCD09801614
  • Molecular Weight : 286.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5206-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5206-34
Availability details for
Quantity
  • Availability
  • Add to Cart