RIE156320894 Rieke Metals, Inc.

3,5-Dichloro-3'-fluoro-4'-methoxybenzophenone

  • MDL number MFCD09801615
  • Molecular Weight : 299.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5206-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5206-48
Availability details for
Quantity
  • Availability
  • Add to Cart