RIE156320896 Rieke Metals, Inc.

3-Chloro-3'-fluoro-4'-methylbenzophenone

  • MDL number MFCD09801617
  • Molecular Weight : 248.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5207-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5207-15
Availability details for
Quantity
  • Availability
  • Add to Cart