RIE156320898 Rieke Metals, Inc.

2-Bromo-3'-fluoro-4'-methylbenzophenone

  • MDL number MFCD09801619
  • Molecular Weight : 293.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5207-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5207-17
Availability details for
Quantity
  • Availability
  • Add to Cart