RIE156320904 Rieke Metals, Inc.

3,3',5-Trifluoro-4'-methylbenzophenone

  • MDL number MFCD09801625
  • Molecular Weight : 250.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5207-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5207-42
Availability details for
Quantity
  • Availability
  • Add to Cart