RIE156320906 Rieke Metals, Inc.

3,5-Dichloro-3'-fluoro-4'-methylbenzophenone

  • MDL number MFCD09801627
  • Molecular Weight : 283.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5207-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5207-48
Availability details for
Quantity
  • Availability
  • Add to Cart