RIE156320907 Rieke Metals, Inc.

2-Chloro-3'-fluoro-5'-methylbenzophenone

  • MDL number MFCD09801628
  • Molecular Weight : 248.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5208-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-14
Availability details for
Quantity
  • Availability
  • Add to Cart