RIE156320908 Rieke Metals, Inc.

3-Chloro-3'-fluoro-5'-methylbenzophenone

  • MDL number MFCD09801629
  • Molecular Weight : 248.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)(3-fluoro-5-methylphenyl)methanone
Inchi Code 1S/C14H10ClFO/c1-9-5-11(8-13(16)6-9)14(17)10-3-2-4-12(15)7-10/h2-8H,1H3
Inchi Key VBSJSSKSGVJLFF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5208-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-15
Availability details for
Quantity
  • Availability
  • Add to Cart