RIE156320909 Rieke Metals, Inc.

4-Chloro-3'-fluoro-5'-methylbenzophenone

  • MDL number MFCD09801630
  • Molecular Weight : 248.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5208-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-16
Availability details for
Quantity
  • Availability
  • Add to Cart