RIE156320912 Rieke Metals, Inc.

4-Bromo-3'-fluoro-5'-methylbenzophenone

  • MDL number MFCD09801633
  • Molecular Weight : 293.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(3-fluoro-5-methylphenyl)methanone
Inchi Code 1S/C14H10BrFO/c1-9-6-11(8-13(16)7-9)14(17)10-2-4-12(15)5-3-10/h2-8H,1H3
Inchi Key QKLWBTULIRZEJY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5208-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-19
Availability details for
Quantity
  • Availability
  • Add to Cart