RIE156320913 Rieke Metals, Inc.

3'-Fluoro-2-iodo-5'-methylbenzophenone

  • MDL number MFCD09801634
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluoro-5-methylphenyl)(2-iodophenyl)methanone
Inchi Code 1S/C14H10FIO/c1-9-6-10(8-11(15)7-9)14(17)12-4-2-3-5-13(12)16/h2-8H,1H3
Inchi Key YJOXYVOROLFEAT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5208-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-20
Availability details for
Quantity
  • Availability
  • Add to Cart