RIE156320914 Rieke Metals, Inc.

3'-Fluoro-3-iodo-5'-methylbenzophenone

  • MDL number MFCD09801635
  • Molecular Weight : 340.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluoro-5-methylphenyl)(3-iodophenyl)methanone
Inchi Code 1S/C14H10FIO/c1-9-5-11(7-12(15)6-9)14(17)10-3-2-4-13(16)8-10/h2-8H,1H3
Inchi Key YSMJHKRMIXSXAS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5208-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-21
Availability details for
Quantity
  • Availability
  • Add to Cart