RIE156320917 Rieke Metals, Inc.

3,3',4-Trifluoro-5'-methylbenzophenone

  • MDL number MFCD09801638
  • Molecular Weight : 250.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5208-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-41
Availability details for
Quantity
  • Availability
  • Add to Cart