RIE156320918 Rieke Metals, Inc.

3,3',5-Trifluoro-5'-methylbenzophenone

  • MDL number MFCD09801639
  • Molecular Weight : 250.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)(3-fluoro-5-methylphenyl)methanone
Inchi Code 1S/C14H9F3O/c1-8-2-9(4-11(15)3-8)14(18)10-5-12(16)7-13(17)6-10/h2-7H,1H3
Inchi Key SVFWHAFTXACPPC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5208-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5208-42
Availability details for
Quantity
  • Availability
  • Add to Cart